missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals

Product Code. 11426126
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-66-5
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00064440
InChI Key CMIBUZBMZCBCAT-IYBDPMFKSA-N
Synonym 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
PubChem CID 12297760
IUPAC Name (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 166°C to 171°C
Beilstein 2022481
Formula Weight 386.36
Percent Purity 98%
Quantity 5 g
Optical Rotation −139° (c=1 in Ethanol)
Chemical Name or Material Di-p-toluoyl-L-tartaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.