Learn More
Thermo Scientific™ dI Phosphoramidite, standard grade, serum vial bottle
dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis.
Brand: Thermo Scientific™ 27-1744-03
11.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis. The problem of degeneracy of the genetic code can be assuaged by utilizing universal bases, such as deoxyinosine (dI), in an oligonucleotide. Its unique structure allows for it to pair with all four bases in various ‘wobble’ structures, with an overall preferential order of base-pairing: dI-dC > dI-dA > dI-dG = dI-dT. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C40H47N6O7P
• Formula weight: 754.8 g/mol
• CAS number: 141684-35-7
• HPLC Purity: ≥98.0%; M-11 Impurity ≤0.01%
• 31P NMR Purity: ≥98%
Specifications
AKTA Synthesizers, Applied Biosystems Synthesizers | |
Store in freezer (at or below -20°C). | |
Room Temperature | |
100 μmol | |
Standard | |
Powder |
dI CEP | |
Deoxyinosine (dI) | |
98% | |
Phosphoramidite | |
DNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.