missing translation for 'onlineSavingsMsg'
Learn More
Dipotassium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10142300

Dipotassium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, Fisher Chemical™

Product Code. 10142300
1 kg
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10142300

Brand: Fisher Chemical P/5245/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number 11383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphateShow MoreShow Less
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340°C
Color White
pH 8.5 to 9.6
Physical Form Solid
Quantity 1 kg
Formula Weight 174.18g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dipotassium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.