missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipotassium Hydrogen Orthophosphate Anhydrous, Extra Pure, SLR, Fisher Chemical™
174.11 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Dipotassium Hydrogen Orthophosphate Anhydrous, Extra Pure, SLR
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 340°C |
Color | White |
pH | 8.5 to 9.6 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 174.18g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'