missing translation for 'onlineSavingsMsg'
Learn More

Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Product Code. 10696070
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34590-94-8
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00059604

Specifications

Melting Point -80.0°C
Density 0.9500g/mL
Boiling Point 180.0°C
Flash Point 75°C
Assay Percent Range 98.5% min. sum of isomers (GC)
Packaging Plastic drum
Linear Formula CH3O(CH2)3O(CH2)3OH
Quantity 5 L
Merck Index 15, 3384
Specific Gravity 0.95
Solubility Information Solubility in water: soluble. Other solubilities: miscible with benzene
Viscosity 4 mPa.s (25°C)
Formula Weight 148.2
Percent Purity 99%
Grade Pure
Chemical Name or Material Di(propylene glycol) methyl ether, Mixture of isomers
Show More Show Less
Product Identifier
  • Di(propylene glycol) methyl ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'