missing translation for 'onlineSavingsMsg'
Learn More
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10132830

Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10132830
5 kg, Plastic keg
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10132830

Brand: Fisher Chemical S/4450/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, Packaging: Plastic powder jar, CAS: 10028-24-7, White, Melting Point: 243 deg.C, Molecular Formula: H5Na2O6P, Molecular Weight: 177.987, pH: 8.7 to 9.4

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2oShow MoreShow Less
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243°C
Color White
pH 8.7 to 9.4
Physical Form Solid
Quantity 5 kg
Formula Weight 177.99g/mol
Packaging Plastic keg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.