missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical, Quantity: 500g, CAS: 10039-32-4, Melting Point: 35 deg.C, Molecular Formula: H25Na2O16P, Molecular Weight: 358.137, Packaging: Plastic powder jar

Specifications
Specifications
Melting Point | 35°C |
pH | 9.0 to 9.4 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 358.13g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
29Hours
15Minutes
17Seconds
36 -
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
8Minutes
42Seconds
23
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction