missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10182730
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number 149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Melting Point 35°C
pH 9.0 to 9.4
Physical Form Solid
Quantity 2.5 kg
Formula Weight 358.13g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.