missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Specifications
Melting Point | 35°C |
pH | 9.0 to 9.4 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 358.13g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'