missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 10519613
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-50-8
Molecular Formula C10H18N2O4
Molecular Weight (g/mol) 230.26
InChI Key QKSQWQOAUQFORH-VAWYXSNFSA-N
Synonym di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester
PubChem CID 6034084
IUPAC Name tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate
SMILES CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C

Specifications

Melting Point 90.0°C to 92.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CO2NNCO2C(CH3)3
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dichloromethane and other chlorinated,solvents
Formula Weight 230.26
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Di-tert-butyl azodicarboxylate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-tert-butyl azodicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'