missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific Chemicals
£29.00 - £323.00
Chemical Identifiers
CAS | 870-50-8 |
---|---|
Molecular Formula | C10H18N2O4 |
Molecular Weight (g/mol) | 230.26 |
InChI Key | QKSQWQOAUQFORH-VAWYXSNFSA-N |
Synonym | di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester |
PubChem CID | 6034084 |
IUPAC Name | tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate |
SMILES | CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10029171
|
Thermo Scientific Acros
366920050 |
5 g | Glass bottle |
£29.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10519613
|
Thermo Scientific Acros
366920250 |
25 g | Glass bottle |
£113.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15790859
|
Thermo Scientific Acros
366921000 |
100 g | Glass bottle |
£323.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiatorsChemical Identifiers
870-50-8 | |
230.26 | |
di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate |
C10H18N2O4 | |
QKSQWQOAUQFORH-VAWYXSNFSA-N | |
6034084 | |
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C |
Specifications
870-50-8 | |
Yellow | |
96% min. (GC) | |
(CH3)3CO2NNCO2C(CH3)3 | |
Solubility in water: insoluble. Other solubilities: soluble in dichloromethane and other chlorinated,solvents | |
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C | |
230.26 | |
230.26 | |
Crystalline Powder or Crystals |
90.0°C to 92.0°C | |
Authentic | |
C10H18N2O4 | |
di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
QKSQWQOAUQFORH-VAWYXSNFSA-N | |
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate | |
6034084 | |
97% | |
Di-tert-butyl azodicarboxylate |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-796-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Di-tert-butyl azodicarboxylate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only