Learn More
Di-tert-butyl dicarbonate, 97+%, Thermo Scientific Chemicals
£24.70 - £232.12
Chemical Identifiers
CAS | 24424-99-5 |
---|---|
Molecular Formula | C10H18O5 |
Molecular Weight (g/mol) | 218.249 |
MDL Number | MFCD00008805 |
InChI Key | DYHSDKLCOJIUFX-UHFFFAOYSA-N |
Synonym | di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonateShow More |
PubChem CID | 90495 |
ChEBI | CHEBI:48500 |
IUPAC Name | tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate |
SMILES | CC(C)(C)OC(=O)OC(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11411950
|
Thermo Scientific Alfa Aesar
A14708.14 |
25 g | N/A | ||||||
11477726
|
Thermo Scientific Alfa Aesar
A14708.14 |
25 g |
£27.38
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487726
|
Thermo Scientific Alfa Aesar
A14708.22 |
100 g | N/A | ||||||
11421950
|
Thermo Scientific Alfa Aesar
A14708.22 |
100 g |
£75.25
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431950
|
Thermo Scientific Alfa Aesar
A14708.36 |
500 g | N/A | ||||||
11497726
|
Thermo Scientific Alfa Aesar
A14708.36 |
500 g |
£232.12
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
Solubility
Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.
Chemical Identifiers
24424-99-5 | |
218.249 | |
DYHSDKLCOJIUFX-UHFFFAOYSA-N | |
90495 | |
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate |
C10H18O5 | |
MFCD00008805 | |
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonateShow More | |
CHEBI:48500 | |
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C |
Specifications
24424-99-5 | |
0.95 | |
37°C (99°F) | |
Mild | |
1.409 | |
UN2930 | |
Moisture sensitive | |
Miscible with decalin,toluene,carbon tetrachloride,tetrahydrofuran,dioxane,alcohols,acetone,acetonitrile and dimethylformamide. Immiscible with water. | |
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C | |
218.249 | |
CHEBI:48500 | |
≥97% |
21°C to 24°C | |
56°C to 57°C (0.5 mmHg) | |
≥97% | |
C10H18O5 | |
MFCD00008805 | |
1911173 | |
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate | |
DYHSDKLCOJIUFX-UHFFFAOYSA-N | |
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate | |
90495 | |
218.25 | |
Di-tert-butyl dicarbonate |
Safety and Handling
GHS H Statement
H330-H318-H228-H315-H317-H335
Fatal if inhaled.
Causes serious eye damage.
Flammable solid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P240-P241-P260-P264b-P271-P272-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P370+P378q-P501c
H228-H315-H317-H318-H330-H335-H500
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 246-240-1
missing translation for 'rtecsNumber' : HT0230000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Moisture sensitive
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.