missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl dicarbonate, 97%

Product Code. 10206680 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10206680

Brand: Thermo Scientific Acros 189775000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Melting Point 22.0°C to 24.0°C
Color White
Density 1.0200g/mL
Boiling Point 56.0°C to 57.0°C (0.5 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O[CO2C(CH3)3]2
Refractive Index 1.4075 to 1.4095
Quantity 500 g
Fieser 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
Merck Index 15, 3038
Specific Gravity 1.02
Formula Weight 218.25
Percent Purity 97%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Di-tert-butyl dicarbonate, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-tert-butyl dicarbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.