missing translation for 'onlineSavingsMsg'
Learn More

(+)-Di-tert-butyl L-tartrate, 99%, Thermo Scientific Chemicals

Product Code. 15462238
Click to view available options
Quantity:
250 mg
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117384-45-9
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.30
MDL Number MFCD00192000
InChI Key ITWOKJQQGHCDBL-UHFFFAOYNA-N
Synonym +-di-tert-butyl l-tartrate, di-tert-butyl l-+-tartrate, 2r,3r-di-tert-butyl 2,3-dihydroxysuccinate, di-t-butyl l-tartrate, r,r-di-tert-butyl-tartrat, di-tert-butyll-+-tartrate, l-tartaric acid di-tert-butyl ester, l-+-tartaric acid di-tert-butyl ester, 1,4-di-tert-butyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 9943166
IUPAC Name ditert-butyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CC(C)(C)OC(=O)C(O)C(O)C(=O)OC(C)(C)C

Specifications

Melting Point 90°C to 92°C
Color White
Density 1.143 g/mL
Flash Point 109°C (228°F)
Quantity 500 mg
Beilstein 5026723
Formula Weight 262.3
Percent Purity 99%
Physical Form Solid
Chemical Name or Material (+)-Di-tert-butyl L-tartrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'