Learn More
Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals
£24.50 - £43.70
Chemical Identifiers
CAS | 110-05-4 |
---|---|
Molecular Formula | C8H18O2 |
Molecular Weight (g/mol) | 146.23 |
MDL Number | MFCD00008803 |
InChI Key | LSXWFXONGKSEMY-UHFFFAOYSA-N |
Synonym | di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d |
PubChem CID | 8033 |
SMILES | CC(C)(C)OOC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10778152
|
Thermo Scientific Acros
349930050 |
5 g | Plastic bottle |
£24.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10367071
|
Thermo Scientific Acros
349931000 |
100 g | Plastic bottle |
£25.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607963
|
Thermo Scientific Acros
349932500 |
250 g | Plastic bottle |
£43.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
110-05-4 | |
146.23 | |
LSXWFXONGKSEMY-UHFFFAOYSA-N | |
8033 |
C8H18O2 | |
MFCD00008803 | |
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d | |
CC(C)(C)OOC(C)(C)C |
Specifications
110-05-4 | |
0.8000g/mL | |
6°C | |
0.1% max. Tert-butyl hydroperoxide (GC) | |
(CH3)3COOC(CH3)3 | |
MFCD00008803 | |
0.8 | |
Solubility in water: immiscible. Other solubilities: soluble in most organic solvents | |
CC(C)(C)OOC(C)(C)C | |
146.23 | |
0.9 mPa.s (20°C) | |
99% | |
Di-tert-butyl peroxide |
-30°C | |
109°C to 110°C | |
Authentic | |
C8H18O2 | |
1.3880 to 1.39 | |
15, 3508 | |
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d | |
LSXWFXONGKSEMY-UHFFFAOYSA-N | |
2-tert-butylperoxy-2-methylpropane | |
8033 | |
146.23 | |
Liquid |
Safety and Handling
GHS H Statement
Heating may cause a fire.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get me
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-733-6
- Di-tert-butyl peroxide
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Germ cell mutagenicity Category 2
- Organic peroxide Type E & F
Hazard Statement
- H341-Suspected of causing genetic defects.
- H412-Harmful to aquatic life with long lasting effects.
- H242-Heating may cause a fire.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only