missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylphenylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Product Code. 15497625
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612088-55-8
Molecular Formula C14H24BF4P
Molecular Weight (g/mol) 310.12
MDL Number MFCD08704553
InChI Key HRDPEVWZXUWEFR-UHFFFAOYSA-O
Synonym di-tert-butylphenylphosphonium tetrafluoroborate, di-tert-butyl phenyl phosphonium tetrafluoroborate, di-tert-butyl phenyl phosphanium tetrafluoroborate, acmc-20al46, di tert-butyl phenylphosphonium tetrafluoroborate
PubChem CID 11220595
IUPAC Name ditert-butyl(phenyl)phosphanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC(C)(C)[PH+](C1=CC=CC=C1)C(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 310.12
Percent Purity 99%
Chemical Name or Material Di-tert-butylphenylphosphonium tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'