missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacetin, 50%, tech. grade, remainder triacetin and monoacetin, Thermo Scientific Chemicals
£15.90 - £119.00
Specifications
Chemical Name or Material | Diacetin |
---|---|
Name Note | pract., 98% |
CAS | 25395-31-7, 102-76-1, 26446-35-5 |
Color | Colorless |
Density | 1.1875g/mL |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10685481
|
Thermo Scientific Acros
406300050 |
5 g | Glass bottle |
£15.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157130
|
Thermo Scientific Acros
406305000 |
500 g | Glass bottle |
£119.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diacetin | |
25395-31-7, 102-76-1, 26446-35-5 | |
1.1875g/mL | |
141°C | |
45% min. (GC) | |
C7H12O5 | |
1.4400 to 1.4500 | |
542 to 605mg KOH/g | |
1.1875 | |
176.17 | |
50% | |
Liquid |
pract., 98% | |
Colorless | |
280.0°C | |
Authentic | |
Glass bottle | |
(CH3COO)2C3H5OH | |
MFCD00008717 | |
15, 2964 | |
Solubility in water: soluble | |
176.17 | |
24 to 28% monoacetin, 41 to 47% diacetin, 22 to 28% triacetin (typically) |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 246-941-2
missing translation for 'rtecsNumber' : AK3325000
missing translation for 'tsca' : TSCA
RUO – Research Use Only