missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 1.19 |
Boiling Point | 250°C to 280°C (decomposition) |
Flash Point | 141°C (285°F) |
Refractive Index | 1.444 |
Quantity | 250 g |
Beilstein | 1706903 |
Sensitivity | Hygroscopic |
Merck Index | 14,2961 |
Solubility Information | Fully miscible in water. |
Formula Weight | 176.17 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction