Learn More
Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals
Description
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.
Solubility
Solubility in hot water gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 107°C to 110°C |
Quantity | 25 g |
Beilstein | 84386 |
Merck Index | 14,2965 |
Optical Rotation | −11° (c=5 in Ethanol) |
Formula Weight | 260.29 |
Percent Purity | ≥98% |
Chemical Name or Material | Diacetone-D-glucose |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252947 | Certificate of Analysis | 11/11/2024 | A15568.22,A15568.14,A15568.36 |
10250298 | Certificate of Analysis | 04/04/2024 | A15568.22,A15568.14,A15568.36 |
10209669 | Certificate of Analysis | 20/07/2023 | A15568,A15568.22,A15568.14,A15568.36 |
10207111 | Certificate of Analysis | 13/07/2022 | A15568.22,A15568.14,A15568.36 |
10138393 | Certificate of Analysis | 23/11/2021 | A15568.22,A15568.14,A15568.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.