missing translation for 'onlineSavingsMsg'
Learn More
Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals Product Code.: 11413037

Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals

Product Code. 11413037
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11413037

Brand: Thermo Scientific Alfa Aesar A15568.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

Solubility
Solubility in hot water gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-52-5
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00005544
InChI Key KEJGAYKWRDILTF-MQIGXGKASA-N
Synonym 1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
PubChem CID 40469256
IUPAC Name (3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 107°C to 110°C
Quantity 100 g
Beilstein 84386
Merck Index 14,2965
Optical Rotation −11° (c=5 in Ethanol)
Formula Weight 260.29
Percent Purity ≥98%
Chemical Name or Material Diacetone-D-glucose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.