missing translation for 'onlineSavingsMsg'
Learn More
Diacetoxydimethylsilane, 95%, AcroSeal™, Thermo Scientific™ Product Code.: 11955931

Diacetoxydimethylsilane, 95%, AcroSeal™, Thermo Scientific™

Product Code. 11955931
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 11955931

Brand: Thermo Scientific Acros 434011000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2182-66-3
Melting Point -12.5°C
Density 1.0530g/mL
Boiling Point 164°C
Flash Point 69°C
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H12O4Si
Linear Formula (CH3COO)2Si(CH3)2
MDL Number MFCD00039832
Quantity 100 mL
Specific Gravity 1.053
Synonym dimethyldiacetoxysilane, diacetoxydimethylsilane, dimethylsilanediyl diacetate, silanediol, dimethyl-, diacetate, dimethylsilanediol, diacetate, silanediol, 1,1-dimethyl-, 1,1-diacetate, acetyloxy dimethylsilyl acetate, dimethyldiacetoxysilicane, dimethyl-silanediodiacetate, acmc-1ak12
InChI Key RQVFGTYFBUVGOP-UHFFFAOYSA-N
SMILES CC(=O)O[Si](C)(C)OC(=O)C
IUPAC Name [acetyloxy(dimethyl)silyl] acetate
Molecular Weight (g/mol) 176.24
PubChem CID 75130
Formula Weight 176.24
Percent Purity 95%
Chemical Name or Material Diacetoxydimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0316078 Certificate TypeCertificate of Analysis Date07/02/2022 Catalog Number
compliance-icons
Product Identifier
  • Diacetoxydimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diacetoxydimethylsilane, 95%, AcroSeal™, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.