missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Diagnostic Kit for Thyroxine (T4) (MPQuanti™ only)
Shop All MP Biomedicals Products
Click to view available options
:
T4
Description
- Diagnostic Kit forThyroxine (T4) (Serum/Plasma) is based on fluorescence immunoassay technology.
- The sample moves through the strip from sample pad to absorbent pad. T4 in the sample will compete with the T4 antigen coated on the membrane.
- The less T4 in the sample, the more fluorescent microspheres conjugated with anti-T4 antibodies can be captured by the T4 antigen coated on the membrane (Test line).
- The concentration of T4 in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
- According to the fluorescence intensity of the test and the standard curve, the concentration of T4 in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show T4 concentration in the sample.
- CE marked
- For professional in-vitro diagnostic use
- Working only with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 87 ̴ 300nmol/L, R≥0.990
- Test results should be interpreted at 15 minutes
Spécification
Spécification
| Detectable Analytes | Thyroxine (T4) |
| Detection Method | Immunofluorescence |
| Sample Type | Human serum, Human plasma |
| Type | POC, Quantitative, T4, thryroxine |
| Form | Ready-to-Use (RTU) |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu