Learn More
Diaion™ CR20, chelating resin, highly porous type, 0.4 mmole/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046781.A1
144.40 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Transitional metal removal without alkali earth metal adsorption
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diaion™ CR20, chelating resin | |
Highly porous type, 0.4mmole/mL on poly(styrene-divinylbenzene) | |
300 to 1180μm | |
1 kg |
300-1180μm Beads | |
Odorless | |
Yellow |
RUO – Research Use Only