missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ CRB05(OH), chelating resin, highly porous type, 0.95 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Boron, arsenic, gallium, and selenium removal
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | 300-850μm Beads |
CAS | 71799-50-3 |
Particle Size | 300 to 850μm |
Density | 1.1 |
Quantity | 1 kg |
Chemical Name or Material | Diaion™ CRB05 (OH), chelating resin, Highly porous type, 0.95meq/mL on poly(styrene-divinylbenzene) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'