missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ HP2MGL, synthetic adsorbent resin, highly porous type, 10g/L on polymethacrylate, P.R. 240 angstroms, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Amino acids separations, small molecules with high hydrophobicity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | 300μm Beads,S.A. 570m |
CAS | 25777-71-3 |
Odor | Odorless |
Particle Size | 300μm, P.R. 240Å |
Molecular Formula | C15H22O6 |
Density | 1.09 g/mL |
Color | White to Yellow |
Synonym | bio acrylic resin, methyl 2-methylprop-2-enoate; 2-2-methylprop-2-enoyloxy ethyl 2-methylprop-2-enoate, unii-eg97988m5q, 2-propenoic acid, 2-methyl-, 1,2-ethanediyl ester, polymer with methyl 2-methyl-2-propenoate, methyl methacrylate, ethylene glycol dimethacrylate polymer, methyl methacrylate, polymer with ethylene glycol dimethacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,2-ethanediyl ester, polymer with methyl 2-methyl-2-propenoate, glycol dimethacrylate; methyl methacrylate, poly methyl methacrylate-co-ethylene glycol dimethacrylate |
Quantity | 1 kg |
InChI Key | XBDAKYZJVVYINU-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'