Learn More
Diaion™ PA412(Cl), ion exchange resin, 6% cross-linked, strongly basic porous type II, 1.1 meq/ml on PS-DVB, Thermo Scientific™
Pure water, nucleic acids and amino acids
Marca: Thermo Scientific Alfa Aesar 046984.A1
Code : NEW
Detalles adicionales : Weight : 1.00000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
300-1180μm Beads | |
6% cross-linked, strongly basic porous type II, 1.1meq/mL on PS-DVB | |
1 kg |
Diaion™ PA412 (Cl), ion exchange resin | |
300 to 1180μm |