missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ PA418(Cl), ion exchange resin, 9% cross-linked, strongly basic porous type II, 1.3 meq/ml on PS-DVB, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Pure water, nucleic acids and amino acids. High resistance to oxidative attack
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | 300-1180μm Beads |
CAS | 69011-15-0 |
Odor | Odorless |
Particle Size | 300 to 1180μm |
Color | Yellow |
Quantity | 1 kg |
Chemical Name or Material | Diaion™ PA418 (Cl), ion exchange resin, 9% cross-linked, strongly basic porous type II, 1.3meq/mL on PS-DVB |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'