missing translation for 'onlineSavingsMsg'
Learn More

Diaion™ SK104(Na), ion exchange resin, 4% cross-linked, strongly acidic gel type, 1.2 meq/ml on PS-DVB, Thermo Scientific™

Specialty process and amino acid separations

Brand:  Thermo Scientific Alfa Aesar 046452.A1

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 15464045

  • £60.70 / 1kg
Estimated Shipment: 27-08-2024
to see stock.

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

300-1180μm Beads
4% cross-linked, strongly acidic gel type, 1.2meq/mL on PS-DVB
300 to 1180μm
Diaion™ SK104 (Na), ion exchange resin
Odorless
1 kg
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers