Learn More
Diallyl phthalate, 98%, Thermo Scientific Chemicals
£20.70 - £104.00
Chemical Identifiers
CAS | 131-17-9 |
---|---|
Molecular Formula | C14H14O4 |
Molecular Weight (g/mol) | 246.26 |
MDL Number | MFCD00008646 |
InChI Key | QUDWYFHPNIMBFC-UHFFFAOYSA-N |
Synonym | diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st |
PubChem CID | 8560 |
IUPAC Name | bis(prop-2-enyl) benzene-1,2-dicarboxylate |
SMILES | C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10559453
|
Thermo Scientific Acros
276622500 |
250 mL | Glass bottle |
£20.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741721
|
Thermo Scientific Acros
276620010 |
1 L | Glass bottle |
£51.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434935
|
Thermo Scientific Acros
276620025 |
2.5 L | Glass bottle |
£104.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131-17-9 | |
246.26 | |
QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
8560 | |
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
C14H14O4 | |
MFCD00008646 | |
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
bis(prop-2-enyl) benzene-1,2-dicarboxylate |
Specifications
131-17-9 | |
Colorless to Yellow | |
300.0°C | |
Authentic | |
C14H14O4 | |
MFCD00008646 | |
09, 4120 | |
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
bis(prop-2-enyl) benzene-1,2-dicarboxylate | |
8560 | |
246.26 | |
Liquid |
-70.0°C | |
1.1200g/mL | |
166°C | |
97.5% min. (GC) | |
1.5180 to 1.5200 | |
3082 | |
1.12 | |
Solubility in water: 148mg/l (20°C) | |
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C | |
246.26 | |
13 mPa.s (20°C) | |
98% | |
Diallyl phthalate |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause damage to organs thr
GHS P Statement
Avoid release to the environment.
Do not get in eyes,on skin,or on clothing.
Call a POISON CENTRE or doctor/physician.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-016-3
- Diallyl phthalate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P262-Do not get in eyes, on skin, or on clothing.
- P273-Avoid release to the environment.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only