Learn More
Bel-Art™ SP Scienceware™ Dialysis Bag Clip Holders
Strong support for suspending dialysis bags in beakers or pitchers
Brand:  Bel-Art™ SP Scienceware™ F18237-0000
Product Code. 15585594
  • £32.50 / Pack of 6
Explore more special offers
Description
Description
  • Suspend dialysis bags in large beakers or pitchers up to 0.63cm (0.25 in.) thick with these strong clips
  • Bags slide easily into slot and then holder can be clipped onto the container’s edge
  • Smooth polypropylene surface may be marked with identifying sequence numbers
  • Autoclavable
Specifications
Specifications
Dialysis Bag Clips
Autoclavable
Natural
Non-sterile
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Product Certifications
Ofertas especiales
Ofertas especiales