Learn More
Bochem™ Diamond Pencil with Octahedron Point
The Bochem™ diamond pencil is used for permanent labelling of microscope slides and other glass products in the lab. Etched glass is resistant to humidity and washing.
Brand:  Bochem™ 12621
Product Code. 15610249
  • £82.30 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent labelling of glassware and microscope slides
  • Octahedron point
  • Steel handle
Specifications
Specifications
Steel
Diamond Pencil
Octahedron Point
135 mm
Permanent Marking Glass Microscope Slides and Other Lab Glassware
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bochem™ Diamond Pencil with Octahedron Point > Laboratory Pencil; Length: 135 mm