missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Diamond Sof-Tred™ Dyna-Shield™ Layer with Non-Directional ''Deck Plate'' Pattern
Notrax Diamond Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
£45.10 - £124.00
Specifications
Color | Black/Yellow |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17782118
|
Notrax
419S0023BY |
91 cm | 60 cm |
£45.10
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
17712128
|
Notrax
419S0035BY |
150 cm | 91 cm |
£124.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
17792108
|
Notrax
419C0024BY |
1830 cm | 60 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17722118
|
Notrax
419C0036BY |
1830 cm | 91 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17752118
|
Notrax
419C0048BY |
1830 cm | 122 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Non-directional ''Deck Plate'' pattern allows freedom of movement
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Black/Yellow | |
Microcellular Vinyl | |
Industrial and Commercial Matting |