missing translation for 'onlineSavingsMsg'
Learn More

Notrax Diamond Sof-Tred™ Dyna-Shield™ Layer with Non-Directional ''Deck Plate'' Pattern

Notrax Diamond Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

Brand:  Notrax 419C0024GY

Additional Details : Weight : 2.30000kg

 View more versions of this product

Product Code. 17702118

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Non-directional ''Deck Plate'' pattern allows freedom of movement
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

60 ft.
Industrial and Commercial Matting
60 cm
Gray
Microcellular Vinyl
1830 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers