Learn More
Notrax Diamond Sof-Tred™ Dyna-Shield™ Layer with Non-Directional ''Deck Plate'' Pattern
Notrax Diamond Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
Brand: Notrax 419S0035GY
Additional Details : Weight : 5.30000kg
Description
- Non-directional ''Deck Plate'' pattern allows freedom of movement
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
59 in. | |
Industrial and Commercial Matting | |
91 cm |
Gray | |
Microcellular Vinyl | |
150 cm |