missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diatomaceous earth, Calcined, Honeywell™ Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 500 g |
Packaging | Poly Bottle |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Diatomaceous Earth, Calcined |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction