missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,d]thiophen-2-ylboronic acid, 97%, Thermo Scientific™

£31.10 - £281.00

Chemical Identifiers

CAS 668983-97-9
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.072
MDL Number MFCD01318982
InChI Key CSLSCVHILGCSTE-UHFFFAOYSA-N
Synonym dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
PubChem CID 2794660
IUPAC Name dibenzothiophen-2-ylboronic acid
SMILES B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10462603
View Documents
Thermo Scientific Maybridge
MO01033CB
250 mg Amber Glass Bottle
£31.10
250mg
Please to purchase this item. Need a web account? Register with us today!
10278522
View Documents
Thermo Scientific Maybridge
MO01033DA
1 g Amber Glass Bottle
£88.60
1g
Please to purchase this item. Need a web account? Register with us today!
10336854
View Documents
Thermo Scientific Maybridge
MO01033DE
5 g Amber Glass Bottle
£281.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

668983-97-9
228.072
CSLSCVHILGCSTE-UHFFFAOYSA-N
2794660
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
C12H9BO2S
MFCD01318982
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
dibenzothiophen-2-ylboronic acid

Specifications

668983-97-9
95%
MFCD01318982
CSLSCVHILGCSTE-UHFFFAOYSA-N
dibenzothiophen-2-ylboronic acid
2794660
95%
95.0
C12H9BO2S
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
228.072
228.08
Dibenzo[b, d]thiophen-2-ylboronic acid, May contain varying amounts of anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications