missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzosuberone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
250 g
Description
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.
Solubility
Soluble in water (0.03g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.
Solubility
Soluble in water (0.03g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 32°C to 34°C |
Density | 1.156 |
Boiling Point | 147°C to 149°C (0.3 mmHg) |
Flash Point | 200°C (392°F) |
Refractive Index | 1.634 |
Quantity | 100 g |
Beilstein | 1454355 |
Solubility Information | Soluble in water (0.03g/L at 20°C). |
IUPAC Name | tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one |
Formula Weight | 208.26 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'