missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberone, 97%, Thermo Scientific Chemicals

Product Code. 11448337
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

Solubility
Soluble in water (0.03g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210-35-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003587
InChI Key BMVWCPGVLSILMU-UHFFFAOYSA-N
Synonym dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
PubChem CID 14589
SMILES O=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Melting Point 32°C to 34°C
Density 1.156
Boiling Point 147°C to 149°C (0.3 mmHg)
Flash Point 200°C (392°F)
Refractive Index 1.634
Quantity 250 g
Beilstein 1454355
Solubility Information Soluble in water (0.03g/L at 20°C).
IUPAC Name 5,6-dihydrodibenzo[2,1-b
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material Dibenzosuberone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'