Learn More
Dibenzothiophene-2-boronic acid, 97%, Thermo Scientific™
£29.80 - £317.00
Chemical Identifiers
CAS | 668983-97-9 |
---|---|
Molecular Formula | C12H9BO2S |
Molecular Weight (g/mol) | 228.072 |
MDL Number | MFCD01318982 |
InChI Key | CSLSCVHILGCSTE-UHFFFAOYSA-N |
Synonym | dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l |
PubChem CID | 2794660 |
IUPAC Name | dibenzothiophen-2-ylboronic acid |
SMILES | B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
668983-97-9 | |
228.072 | |
CSLSCVHILGCSTE-UHFFFAOYSA-N | |
2794660 | |
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O |
C12H9BO2S | |
MFCD01318982 | |
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l | |
dibenzothiophen-2-ylboronic acid |
Specifications
668983-97-9 | |
C12H9BO2S | |
15124 | |
CSLSCVHILGCSTE-UHFFFAOYSA-N | |
dibenzothiophen-2-ylboronic acid | |
2794660 | |
97% | |
Dibenzothiophene-2-boronic acid |
White | |
MFCD01318982 | |
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l | |
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O | |
228.072 | |
228.07 | |
Powder |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C