missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11403257
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62708-56-9
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149119
InChI Key YONLFQNRGZXBBF-OKILXGFUSA-N
Synonym 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
PubChem CID 44119738
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 90°C to 92°C
Solubility Information Soluble in methanol,acetone and organic solvents. Slightly soluble in water.
IUPAC Name (2R,3S)-2,3-bis(benzoyloxy)butanedioic acid
Formula Weight 376.33 (358.31 Anhydrous)
Percent Purity ≥98%
Quantity 2500 g
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.