missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoylmethane, 98%

Product Code. 10408450 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
25g
500g
This item is not returnable. View return policy

Product Code. 10408450

Brand: Thermo Scientific Acros 112600250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-46-7 | C15H12O2 | 224.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 75°C to 79°C
Color Yellow
Boiling Point 219°C to 221°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5COCH2COC6H5
Quantity 25 g
Beilstein 07,769
Merck Index 15,3014
Formula Weight 224.26
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Dibenzoylmethane, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.