missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl carbonate, 98%, Thermo Scientific Chemicals

£56.60 - £767.00

Chemical Identifiers

CAS 3459-92-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00014436
InChI Key PIZLBWGMERQCOC-UHFFFAOYSA-N
Synonym carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
PubChem CID 77002
IUPAC Name dibenzyl carbonate
SMILES O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450343
View Documents
Thermo Scientific Alfa Aesar
A11464.09
10 g
£56.60
10g
Please to purchase this item. Need a web account? Register with us today!
11460343
View Documents
Thermo Scientific Alfa Aesar
A11464.18
50 g
£223.00
50g
Please to purchase this item. Need a web account? Register with us today!
11470343
View Documents
Thermo Scientific Alfa Aesar
A11464.30
250 g
£767.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

Solubility
Sparingly Soluble (0.099 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Chemical Identifiers

3459-92-5
242.27
PIZLBWGMERQCOC-UHFFFAOYSA-N
77002
O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
C15H14O3
MFCD00014436
carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
dibenzyl carbonate

Specifications

3459-92-5
150°C to 155°C (0.8 mmHg)
C15H14O3
MFCD00014436
Air sensitive
Sparingly Soluble (0.099g/L) (25°C).
O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
242.27
242.27
Dibenzyl carbonate
27°C to 30°C
>110°C (230°F)
1.5485
1882864
carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
PIZLBWGMERQCOC-UHFFFAOYSA-N
dibenzyl carbonate
77002
98%
Videos
Safety and Handling

Safety and Handling

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'einecsNumber' : 222-401-1

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only