missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl diisopropylphosphoramidite, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.03 |
Boiling Point | 130°C (0.55 mmHg) |
Flash Point | 69°C (156°F) |
Assay Percent Range | 0.9 |
Quantity | 100 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform. |
Formula Weight | 345.43 |
Percent Purity | 95% |
Chemical Name or Material | Dibenzyl diisopropylphosphoramidite |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'