missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl diselenide, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 91°C to 93°C |
Quantity | 1 g |
UN Number | UN3283 |
Beilstein | 1877745 |
Solubility Information | Soluble in water. |
Formula Weight | 340.18 |
Percent Purity | 95% |
Chemical Name or Material | Dibenzyl diselenide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'