missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyldithiocarbamic acid zinc salt, 95%, Thermo Scientific Chemicals

£87.03 - £270.35

Chemical Identifiers

CAS 14726-36-4
Molecular Formula C30H28N2S4Zn
Molecular Weight (g/mol) 610.21
InChI Key AUMBZPPBWALQRO-UHFFFAOYSA-L
Synonym zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc
PubChem CID 84610
IUPAC Name zinc;N,N-dibenzylcarbamodithioate
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10288580
SDS View Documents
Thermo Scientific Acros
406440250
25 g Glass bottle
£87.03
25g
Please to purchase this item. Need a web account? Register with us today!
11985711
SDS View Documents
Thermo Scientific Acros
406441000
100 g Glass bottle
£270.35
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dibenzyldithiocarbamic acid zinc salt, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.