Learn More
Dibromoacetonitrile, 94%, Thermo Scientific Chemicals
£18.00 - £164.00
Chemical Identifiers
CAS | 3252-43-5 |
---|---|
Molecular Formula | C2HBr2N |
Molecular Weight (g/mol) | 198.845 |
MDL Number | MFCD00001856 |
InChI Key | NDSBDLSWTGLNQA-UHFFFAOYSA-N |
Synonym | dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 |
PubChem CID | 18617 |
ChEBI | CHEBI:82445 |
IUPAC Name | 2,2-dibromoacetonitrile |
SMILES | C(#N)C(Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458836
|
Thermo Scientific Alfa Aesar
A16994.06 |
5 g |
£18.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468836
|
Thermo Scientific Alfa Aesar
A16994.14 |
25 g |
£49.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478836
|
Thermo Scientific Alfa Aesar
A16994.22 |
100 g |
£164.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3252-43-5 | |
198.845 | |
NDSBDLSWTGLNQA-UHFFFAOYSA-N | |
18617 | |
2,2-dibromoacetonitrile |
C2HBr2N | |
MFCD00001856 | |
dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 | |
CHEBI:82445 | |
C(#N)C(Br)Br |
Specifications
3252-43-5 | |
67°C to 69°C (24 mmHg) | |
Pungent, Makes Eyes Water | |
Br2CHCN | |
MFCD00001856 | |
1739037 | |
dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 | |
C(#N)C(Br)Br | |
198.845 | |
CHEBI:82445 | |
94% |
2.296 | |
56°C (133°F) | |
C2HBr2N | |
1.542 | |
UN3286 | |
Light sensitive | |
NDSBDLSWTGLNQA-UHFFFAOYSA-N | |
2,2-dibromoacetonitrile | |
18617 | |
198.84 | |
Dibromoacetonitrile |
Safety and Handling
GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H319-H335-H351
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 221-843-2
missing translation for 'rtecsNumber' : AL8450000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only