missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromomalonamide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
Solubility
Soluble in water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
Solubility
Soluble in water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 202°C to 207°C |
Quantity | 10 g |
Beilstein | 1770873 |
Sensitivity | Light and moisture sensitive |
Synonym | dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide |
Solubility Information | Soluble in water. |
IUPAC Name | dibromopropanediamide |
Formula Weight | 259.89 |
Percent Purity | ≥98% |
Chemical Name or Material | Dibromomalonamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'