missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin dichloride, 96%

Product Code. 10308720 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10308720

Brand: Thermo Scientific Acros 194872500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 683-18-1 | C8H18Cl2Sn | 303.842 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-18-1
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 39.0°C to 41.0°C
Color White to Beige
Density 1.4000g/mL
Boiling Point 135.0°C (10.0 mmHg)
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H18Cl2Sn
Linear Formula [CH3(CH2)3]2SnCl2
Quantity 250 g
Fieser 01,213; 01,1294; 05,622
Specific Gravity 1.4
Molecular Weight (g/mol) 303.842
Formula Weight 303.83
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Dibutyltin dichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibutyltin dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.