Learn More
Dibutyltin oxide, 98%, Thermo Scientific Chemicals
£22.21 - £68.44
Chemical Identifiers
CAS | 818-08-6 |
---|---|
Molecular Formula | C8H18OSn |
Molecular Weight (g/mol) | 248.92 |
MDL Number | MFCD00001992 |
InChI Key | JGFBRKRYDCGYKD-UHFFFAOYSA-N |
Synonym | dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tinShow More |
PubChem CID | 61221 |
IUPAC Name | dibutyl(oxo)tin |
SMILES | CCCC[Sn](=O)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10513741
|
Thermo Scientific
179361000 |
100 g | Plastic bottle |
£22.21
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10194380
|
Thermo Scientific Acros
179365000 |
500 g | Plastic bottle |
£68.44
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
818-08-6 | |
248.92 | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
61221 | |
CCCC[Sn](=O)CCCC |
C8H18OSn | |
MFCD00001992 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tinShow More | |
dibutyl(oxo)tin |
Specifications
818-08-6 | |
White | |
98% | |
[CH3(CH2)3]2Sn(=O) | |
04, I, 588 | |
15, 3048 | |
Solubility in water: soluble. Other solubilities: practically insoluble in organic solvents | |
CCCC[Sn](=O)CCCC | |
248.92 | |
248.92 | |
Powder or Granular Powder |
>300.0°C | |
Authentic | |
C8H18OSn | |
MFCD00001992 | |
05,188; 09,141; 10,123; 12,160; 13,53; 15,116; 16,112 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
dibutyl(oxo)tin | |
61221 | |
98% | |
Dibutyltin oxide, 98% |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-449-1
missing translation for 'rtecsNumber' : WH7175000
missing translation for 'tsca' : TSCA
- Dibutyltin oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360-May damage fertility or the unborn child.
- H341-Suspected of causing genetic defects.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.