missing translation for 'onlineSavingsMsg'
Learn More
Dicarbonylbis(cyclopentadienyl)titanium(II), 98%, Thermo Scientific Chemicals Product Code.: 11384467

Dicarbonylbis(cyclopentadienyl)titanium(II), 98%, Thermo Scientific Chemicals

Product Code. 11384467
2 g
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy

Product Code. 11384467

Brand: Thermo Scientific Alfa Aesar 039571.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12129-51-0
Molecular Formula C12H12O2Ti
Molecular Weight (g/mol) 236.09
MDL Number MFCD00143762
InChI Key AGUCYLATRKJGHM-UHFFFAOYSA-N
Synonym dicarbonylbis cyclopentadienyl titanium ii, acmc-1bra9, bis oxomethylidene titanium;cyclopenta-1,3-diene

Specifications

Melting Point 90°C (decomposition)
Boiling Point 60°C (0.001 mmHg)(sublimation)
Quantity 2 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 234.09
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Dicarbonylbis(cyclopentadienyl)titanium(II)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU17J044 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number039571.02,039571.04
Lot NumberH10S008 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number039571.02,039571.04
Lot NumberC18Q17 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number039571.02,039571.04
Lot NumberU09E010 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number039571.02,039571.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicarbonylbis(cyclopentadienyl)titanium(II), 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.